Assay Details
| Gene Symbol: Spast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162870.1 | NP_001156342.1 | 11 - 12 | 1456 | 82 | |
| NM_016962.2 | NP_058658.2 | 11 - 12 | 1453 | 82 | ||
| GenBank mRNA | AJ246002.1 | - | 11 - 12 | 1078 | 82 | |
| AK007793.1 | - | 11 - 12 | 1233 | 82 | ||
| AK044900.1 | - | 11 - 12 | 1375 | 82 | ||
| AK129282.1 | - | 11 - 12 | 1430 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spast
Genomic Map