Assay Details
| Gene Symbol: Spast | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162870.1 | NP_001156342.1 | 13 - 14 | 1584 | 107 | |
| NM_016962.2 | NP_058658.2 | 13 - 14 | 1581 | 107 | ||
| GenBank mRNA | AJ246002.1 | - | 13 - 14 | 1206 | 107 | |
| AK007793.1 | - | 13 - 14 | 1361 | 107 | ||
| AK129282.1 | - | 13 - 14 | 1558 | 107 | ||
| BC046286.1 | - | 13 - 14 | 1581 | 6441742 | 107 | |
More Information
Set Membership: |
|

Spast
Genomic Map