| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700010868.mm.1 |
NM_001162870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010868.mm.1 |
NM_001162870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010868.mm.1 |
NM_016962.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010868.mm.1 |
NM_016962.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010868.mm.1 |
TC1700001156.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700010870.mm.1 |
NM_001162870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010870.mm.1 |
NM_001162870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010870.mm.1 |
NM_016962.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010870.mm.1 |
NM_016962.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010870.mm.1 |
TC1700001156.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700010871.mm.1 |
NM_001162870.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010871.mm.1 |
NM_001162870.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010871.mm.1 |
NM_016962.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010871.mm.1 |
NM_016962.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700010871.mm.1 |
TC1700001156.mm.1 |
MTA TC |