Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 12 - 13 | 1699 | 91 | |
| GenBank mRNA | AF203969.1 | - | 12 - 13 | 1552 | 91 | |
| AK010566.1 | - | 5 - 6 | 766 | 91 | ||
| AK045238.1 | - | 1 - 2 | 69 | 91 | ||
| AK149953.1 | - | 12 - 13 | 1626 | 91 | ||
| AK157205.1 | - | 12 - 13 | 1641 | 91 | ||
| AK158036.1 | - | 12 - 13 | 1612 | 91 | ||
| AK170842.1 | - | 12 - 13 | 1584 | 91 | ||
| BC006748.1 | - | 7 - 8 | 1254 | 91 | ||
| BC023297.1 | - | 12 - 13 | 1585 | 4192224 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map