Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 13 - 14 | 1796 | 87 | |
| GenBank mRNA | AK010566.1 | - | 6 - 7 | 863 | 87 | |
| AK045238.1 | - | 2 - 3 | 166 | 87 | ||
| AK149953.1 | - | 13 - 14 | 1723 | 87 | ||
| AK157205.1 | - | 13 - 14 | 1738 | 87 | ||
| AK170842.1 | - | 13 - 14 | 1681 | 87 | ||
| AK171838.1 | - | 13 - 14 | 1727 | 87 | ||
| BC006748.1 | - | 8 - 9 | 1351 | 87 | ||
| BC023297.1 | - | 13 - 14 | 1682 | 4192224 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map