Assay Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018813.3 | NP_061283.2 | 2 - 3 | 307 | 113 | |
| GenBank mRNA | AF203969.1 | - | 2 - 3 | 160 | 113 | |
| AK149953.1 | - | 2 - 3 | 234 | 113 | ||
| AK157205.1 | - | 2 - 3 | 249 | 113 | ||
| AK158036.1 | - | 2 - 3 | 219 | 113 | ||
| AK170842.1 | - | 2 - 3 | 192 | 113 | ||
| AK171838.1 | - | 2 - 3 | 238 | 113 | ||
| BC023297.1 | - | 2 - 3 | 193 | 4192224 | 113 | |
| BY094575.1 | - | 2 - 3 | 311 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3
Genomic Map