Assay Details
| Gene Symbol: Rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009044.2 | NP_033070.2 | 7 - 8 | 1139 | 100 | |
| GenBank mRNA | AK031627.1 | - | 7 - 8 | 937 | 100 | |
| BC139447.1 | - | 7 - 8 | 1079 | 8862469 | 100 | |
| BC139448.1 | - | 7 - 8 | 1079 | 8862470 | 100 | |
| BC139770.1 | - | 7 - 8 | 1081 | 40111461 | 100 | |
| X60271.1 | - | 7 - 8 | 1138 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rel
Genomic Map