Assay Details
| Gene Symbol: Papolg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172555.2 | NP_766143.2 | 2 - 3 | 411 | 148 | |
| GenBank mRNA | AK035810.1 | - | 2 - 3 | 394 | 148 | |
| AK042629.1 | - | 2 - 3 | 378 | 148 | ||
| AK139065.1 | - | 2 - 3 | 378 | 148 | ||
| BC049163.1 | - | 2 - 3 | 357 | 148 | ||
| BC059271.1 | - | 2 - 3 | 405 | 2811338 | 148 | |
| DQ103505.1 | - | 2 - 3 | 217 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papolg
Genomic Map