Assay Details
| Gene Symbol: Papolg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172555.2 | NP_766143.2 | 4 - 5 | 560 | 143 | |
| GenBank mRNA | AK035810.1 | - | 4 - 5 | 543 | 143 | |
| BC049163.1 | - | 4 - 5 | 506 | 143 | ||
| BC059271.1 | - | 4 - 5 | 554 | 2811338 | 143 | |
| DQ103505.1 | - | 4 - 5 | 366 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papolg
Genomic Map