Assay Details
| Gene Symbol: Mybl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290397.1 | NP_001277326.1 | 1 - 2 | 276 | 147 | |
| NM_008651.3 | NP_032677.2 | 1 - 2 | 276 | 147 | ||
| GenBank mRNA | L35261.1 | - | 1 - 2 | 275 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L35261.1, NM_001290397.1, NM_008651.3 transcripts.
Set Membership: |
|

Mybl1
Genomic Map