Assay Details
| Gene Symbol: Mybl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290397.1 | NP_001277326.1 | 7 - 8 | 1011 | 109 | |
| NM_008651.3 | NP_032677.2 | 7 - 8 | 1011 | 109 | ||
| GenBank mRNA | BC101945.1 | - | 7 - 8 | 755 | 109 | |
| BC103533.1 | - | 7 - 8 | 756 | 40039678 | 109 | |
| BC103534.1 | - | 7 - 8 | 755 | 109 | ||
| L35261.1 | - | 7 - 8 | 1010 | 109 | ||
| X82327.1 | - | 7 - 8 | 756 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybl1
Genomic Map