Assay Details
| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024448.2 | NP_077768.2 | 4 - 5 | 611 | 60 | |
| GenBank mRNA | AB232607.1 | - | 4 - 5 | 511 | 60 | |
| AK013690.1 | - | 3 - 4 | 267 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab12
Genomic Map