Assay Details
Gene Symbol: Themis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028998.1 | NP_083274.1 | 1 - 2 | 175 | 80 | |
GenBank mRNA | AK018663.1 | - | 1 - 2 | 176 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
