Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700022576.mm.1 |
NM_028998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022576.mm.1 |
NM_028998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022576.mm.1 |
TC1700002437.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022577.mm.1 |
NM_028998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022577.mm.1 |
NM_028998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022577.mm.1 |
TC1700002437.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022579.mm.1 |
NM_028998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022579.mm.1 |
NM_028998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022579.mm.1 |
TC1700002437.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700022578.mm.1 |
NM_028998.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022578.mm.1 |
NM_028998.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700022578.mm.1 |
TC1700002437.mm.1 |
MTA TC |