Assay Details
| Gene Symbol: Wash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037757.1 | NP_001032846.1 | 6 - 7 | 769 | 76 | |
| NM_026833.1 | NP_081109.1 | 6 - 7 | 786 | 76 | ||
| GenBank mRNA | AJ304796.1 | - | 6 - 7 | 718 | 76 | |
| AK033616.1 | - | 6 - 7 | 763 | 76 | ||
| AK149967.1 | - | 6 - 7 | 702 | 76 | ||
| AK154400.1 | - | 6 - 7 | 771 | 76 | ||
| AK159820.1 | - | 6 - 7 | 714 | 76 | ||
| AK171737.1 | - | 6 - 7 | 704 | 76 | ||
| AK172673.1 | - | 6 - 7 | 704 | 76 | ||
| BC025839.1 | - | 6 - 7 | 699 | 5136195 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wash1
Genomic Map