Assay Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252192.1 | NP_001239121.1 | 10 - 11 | 1529 | 59 | |
| NM_010164.2 | NP_034294.2 | 11 - 12 | 1645 | 59 | ||
| GenBank mRNA | AK042412.1 | - | 10 - 11 | 1452 | 59 | |
| AK153788.1 | - | 11 - 12 | 1651 | 59 | ||
| AK162111.1 | - | 3 - 4 | 386 | 59 | ||
| BC060260.1 | - | 11 - 12 | 1645 | 6848408 | 59 | |
| BC066860.1 | - | 11 - 12 | 1299 | 30089692 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eya1
Genomic Map