Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023963.mm.1 |
NM_025485.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023963.mm.1 |
NM_025485.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023963.mm.1 |
TC0900002908.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023961.mm.1 |
NM_025485.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023961.mm.1 |
NM_025485.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023961.mm.1 |
TC0900002908.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023962.mm.1 |
NM_025485.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023962.mm.1 |
NM_025485.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023962.mm.1 |
TC0900002908.mm.1 |
MTA TC |