Assay Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015827.2 | NP_056642.1 | 15 - 16 | 1976 | 67 | |
| GenBank mRNA | AF043120.1 | - | 4 - 5 | 521 | 67 | |
| AK088064.1 | - | 15 - 16 | 1977 | 67 | ||
| AK153352.1 | - | 15 - 16 | 1977 | 67 | ||
| BC006675.1 | - | 15 - 16 | 1963 | 3499585 | 67 | |
| BC010842.1 | - | 15 - 16 | 1965 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copb2
Genomic Map