Assay Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015827.2 | NP_056642.1 | 16 - 17 | 2084 | 104 | |
| GenBank mRNA | AF043120.1 | - | 5 - 6 | 629 | 104 | |
| AK088064.1 | - | 16 - 17 | 2085 | 104 | ||
| AK153352.1 | - | 16 - 17 | 2085 | 104 | ||
| BC006675.1 | - | 16 - 17 | 2071 | 3499585 | 104 | |
| BC010842.1 | - | 16 - 17 | 2073 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copb2
Genomic Map