Assay Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015827.2 | NP_056642.1 | 7 - 8 | 839 | 67 | |
| GenBank mRNA | AK088064.1 | - | 7 - 8 | 840 | 67 | |
| AK153352.1 | - | 7 - 8 | 840 | 67 | ||
| BC006675.1 | - | 7 - 8 | 826 | 3499585 | 67 | |
| BC010842.1 | - | 7 - 8 | 829 | 67 | ||
More Information
Set Membership: |
|

Copb2
Genomic Map