Assay Details
| Gene Symbol: Rbm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172762.2 | NP_766350.2 | 1 - 2 | 96 | 114 | |
| GenBank mRNA | AK038729.1 | - | 1 - 2 | 67 | 114 | |
| AK078099.1 | - | 1 - 2 | 89 | 114 | ||
| AK079563.1 | - | 1 - 2 | 67 | 114 | ||
| AK136464.1 | - | 1 - 2 | 67 | 114 | ||
| BB862436.1 | - | 1 - 2 | 141 | 114 | ||
| BC141278.1 | - | 1 - 2 | 85 | 9055890 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm34
Genomic Map