Assay Details
| Gene Symbol: Nelfcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020580.3 | NP_065605.2 | 6 - 7 | 750 | 116 | |
| GenBank mRNA | AJ238741.1 | - | 6 - 7 | 684 | 116 | |
| AJ238742.1 | - | 6 - 7 | 750 | 116 | ||
| AJ238743.1 | - | 6 - 7 | 704 | 116 | ||
| AK010360.1 | - | 6 - 7 | 697 | 116 | ||
| AK049418.1 | - | 6 - 7 | 722 | 116 | ||
| AK159866.1 | - | 6 - 7 | 751 | 116 | ||
| BC006959.1 | - | 6 - 7 | 711 | 3707929 | 116 | |
| BC031747.1 | - | 6 - 7 | 708 | 3673038 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nelfcd
Genomic Map