Assay Details
| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008652.2 | NP_032678.1 | 2 - 3 | 310 | 93 | |
| GenBank mRNA | AK028497.1 | - | 2 - 3 | 310 | 93 | |
| BC050842.1 | - | 2 - 3 | 256 | 6518298 | 93 | |
| X70472.1 | - | 2 - 3 | 258 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybl2
Genomic Map