Assay Details
| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008652.2 | NP_032678.1 | 5 - 6 | 690 | 77 | |
| GenBank mRNA | AK028497.1 | - | 5 - 6 | 690 | 77 | |
| AK183670.1 | - | 2 - 3 | 283 | 77 | ||
| BC050842.1 | - | 5 - 6 | 636 | 6518298 | 77 | |
| X70472.1 | - | 5 - 6 | 638 | 77 | ||
More Information
Set Membership: |
|

Mybl2
Genomic Map