Assay Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175646.3 | NP_783577.2 | 1 - 2 | 76 | 64 | |
| GenBank mRNA | AK079533.1 | - | 1 - 2 | 174 | 64 | |
| BC094379.1 | - | 1 - 2 | 92 | 30541506 | 64 | |
| BY205801.1 | - | 1 - 2 | 70 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC094379.1,NM_175646.3 transcripts.
This assay is in the 5'UTR region of the BC094379.1,NM_175646.3 transcripts.
Set Membership: |
|

Txnl4b
Genomic Map