Assay Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028018.2 | NP_082294.1 | 5 - 6 | 572 | 127 | |
| GenBank mRNA | AK010262.1 | - | 5 - 6 | 542 | 127 | |
| AK050468.1 | - | 5 - 6 | 539 | 127 | ||
| AK088192.1 | - | 5 - 6 | 527 | 127 | ||
| AK122215.1 | - | 3 - 4 | 346 | 127 | ||
| AK152830.1 | - | 5 - 6 | 539 | 127 | ||
| AK161899.1 | - | 5 - 6 | 528 | 127 | ||
| AK207302.1 | - | 3 - 4 | 266 | 127 | ||
| BC017605.1 | - | 5 - 6 | 538 | 4507347 | 127 | |
| BC039052.1 | - | 5 - 6 | 515 | 2631157 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ist1
Genomic Map