Assay Details
| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011998.4 | NP_036128.3 | 1 - 2 | 94 | 68 | |
| GenBank mRNA | AK009113.1 | - | 1 - 2 | 95 | 68 | |
| BC057886.1 | - | 1 - 2 | 114 | 4987327 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009113.1, BC057886.1, NM_011998.4 transcripts.
Set Membership: |
|

Chst4
Genomic Map