Assay Details
| Gene Symbol: Stac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016853.2 | NP_058549.1 | 7 - 8 | 945 | 82 | |
| GenBank mRNA | AK134777.1 | - | 6 - 7 | 675 | 82 | |
| AK143440.1 | - | 7 - 8 | 978 | 82 | ||
| BC107325.1 | - | 7 - 8 | 869 | 40054255 | 82 | |
| BC107326.1 | - | 7 - 8 | 869 | 40054256 | 82 | |
| D86639.1 | - | 7 - 8 | 945 | 82 | ||
More Information
Set Membership: |
|

Stac
Genomic Map