Assay Details
| Gene Symbol: Stac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016853.2 | NP_058549.1 | 9 - 10 | 1074 | 80 | |
| GenBank mRNA | AK134777.1 | - | 8 - 9 | 804 | 80 | |
| AK143440.1 | - | 9 - 10 | 1107 | 80 | ||
| BC107325.1 | - | 9 - 10 | 998 | 40054255 | 80 | |
| BC107326.1 | - | 9 - 10 | 998 | 40054256 | 80 | |
| D86639.1 | - | 9 - 10 | 1074 | 80 | ||
More Information
Set Membership: |
|

Stac
Genomic Map