Assay Details
| Gene Symbol: Nudt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024446.5 | NP_077766.3 | 3 - 4 | 387 | 82 | |
| GenBank mRNA | AK011172.1 | - | 3 - 4 | 321 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nudt7
Genomic Map