Assay Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080463.3 | NP_536711.3 | 2 - 3 | 316 | 67 | |
| GenBank mRNA | AF375885.1 | - | 2 - 3 | 285 | 67 | |
| AK037857.1 | - | 2 - 3 | 280 | 67 | ||
| AK044629.1 | - | 2 - 3 | 294 | 67 | ||
| AK081059.1 | - | 2 - 3 | 273 | 67 | ||
| AK132301.1 | - | 2 - 3 | 271 | 67 | ||
| AK135635.1 | - | 2 - 3 | 298 | 67 | ||
| AK170038.1 | - | 2 - 3 | 271 | 67 | ||
| BC046295.1 | - | 2 - 3 | 316 | 6485896 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pofut1
Genomic Map