Assay Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080463.3 | NP_536711.3 | 3 - 4 | 505 | 65 | |
| GenBank mRNA | AF375885.1 | - | 3 - 4 | 474 | 65 | |
| AK044629.1 | - | 3 - 4 | 483 | 65 | ||
| AK081059.1 | - | 3 - 4 | 462 | 65 | ||
| AK132301.1 | - | 3 - 4 | 460 | 65 | ||
| AK135635.1 | - | 3 - 4 | 487 | 65 | ||
| AK170038.1 | - | 3 - 4 | 460 | 65 | ||
| BC046295.1 | - | 3 - 4 | 505 | 6485896 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pofut1
Genomic Map