Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 13 - 14 | 1587 | 86 | |
| GenBank mRNA | AB093220.2 | - | 12 - 13 | 1331 | 86 | |
| AK035622.1 | - | 13 - 14 | 1549 | 86 | ||
| AK048158.1 | - | 13 - 14 | 1345 | 86 | ||
| AK083541.1 | - | 13 - 14 | 1587 | 86 | ||
| AK087048.1 | - | 13 - 14 | 1340 | 86 | ||
| AK088012.1 | - | 13 - 14 | 1343 | 86 | ||
| AK162435.1 | - | 13 - 14 | 1348 | 86 | ||
| AK171487.1 | - | 13 - 14 | 1345 | 86 | ||
| AK210804.1 | - | 4 - 5 | 310 | 86 | ||
| BC110309.1 | - | 13 - 14 | 1343 | 5324218 | 86 | |
| BC116320.2 | - | 13 - 14 | 1539 | 40096002 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map