Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 4 - 5 | 650 | 59 | |
| GenBank mRNA | AB093220.2 | - | 3 - 4 | 394 | 59 | |
| AK035622.1 | - | 4 - 5 | 612 | 59 | ||
| AK047497.1 | - | 4 - 5 | 404 | 59 | ||
| AK048158.1 | - | 4 - 5 | 408 | 59 | ||
| AK083541.1 | - | 4 - 5 | 650 | 59 | ||
| AK087048.1 | - | 4 - 5 | 403 | 59 | ||
| AK088012.1 | - | 4 - 5 | 406 | 59 | ||
| AK162435.1 | - | 4 - 5 | 411 | 59 | ||
| AK171487.1 | - | 4 - 5 | 408 | 59 | ||
| BC110309.1 | - | 4 - 5 | 406 | 5324218 | 59 | |
| BC116320.2 | - | 4 - 5 | 602 | 40096002 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map