Assay Details
| Gene Symbol: Inpp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008916.2 | NP_032942.1 | 9 - 10 | 1245 | 70 | |
| GenBank mRNA | AF483522.1 | - | 9 - 10 | 1165 | 70 | |
| AF483523.1 | - | 9 - 10 | 1165 | 70 | ||
| AK052817.1 | - | 8 - 9 | 1989 | 70 | ||
| AK054436.1 | - | 9 - 10 | 1264 | 70 | ||
| AK078104.1 | - | 9 - 10 | 1256 | 70 | ||
| AK164290.1 | - | 9 - 10 | 1234 | 70 | ||
| BC014725.1 | - | 9 - 10 | 1175 | 3969089 | 70 | |
| BC066112.1 | - | 9 - 10 | 1245 | 30361046 | 70 | |
| U96724.1 | - | 9 - 10 | 1276 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5k
Genomic Map