Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027773.3 | NP_082049.1 | 3 - 4 | 486 | 73 | |
GenBank mRNA | AK077374.1 | - | 3 - 4 | 446 | 73 | |
AK160183.1 | - | 3 - 4 | 447 | 73 | ||
BC115519.1 | - | 3 - 4 | 485 | 40101189 | 73 | |
BC115520.1 | - | 3 - 4 | 486 | 40101190 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
