Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027773.3 | NP_082049.1 | 4 - 5 | 584 | 64 | |
GenBank mRNA | AK077374.1 | - | 4 - 5 | 544 | 64 | |
AK160183.1 | - | 4 - 5 | 545 | 64 | ||
BC115519.1 | - | 4 - 5 | 583 | 40101189 | 64 | |
BC115520.1 | - | 4 - 5 | 584 | 40101190 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930554 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930555 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930552 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930553 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930558 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997310 | AK160183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997310 | BC115519.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997310 | BC115520.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997310 | AK077374.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997310 | NM_027773.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P255849 | AK160183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P255849 | AK077374.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P255849 | AK160183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P255849 | AK077374.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 946196 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566760 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43411 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169029 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173547 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P255849 | AK160183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P255849 | AK077374.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P255849 | AK160183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P255849 | AK077374.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997310 | AK160183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997310 | BC115519.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997310 | BC115520.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997310 | AK077374.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997310 | NM_027773.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P110618 | AK036850.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P110618 | AK030014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P255849 | AK160183.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P255849 | AK077374.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P255849 | AK160183.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P255849 | AK077374.1 | mouse_transcriptome |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930554 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930558 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930553 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930555 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27930552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27930550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930597 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09262265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09223756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08976149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09651473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930591 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09651461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09010710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09037464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09004690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09232799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09277652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09651470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09651469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09340045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27930546 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10378756 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378762 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378758 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378757 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378755 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378760 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10378759 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435076_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10378754 | - | - |
Mouse Genome 430 2.0 Array | 1435076_at | - | - |
Mouse Genome 430 2.0 Array | 1435076_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116972_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 116972_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100009964.mm.1 | NM_027773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100009964.mm.1 | NM_027773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100009964.mm.1 | TC1100001095.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100009963.mm.1 | NM_027773.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100009963.mm.1 | NM_027773.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100009963.mm.1 | TC1100001095.mm.1 | MTA TC |
Mouse Expression Set 430 | 1435076_PM_at | - | - |
Mouse Expression Set 430 | 1435076_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435076_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435076_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240031 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2909105 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2909105 | - | - |
