Assay Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029548.4 | NP_083824.1 | 3 - 4 | 390 | 132 | |
| GenBank mRNA | AB112925.1 | - | 3 - 4 | 132 | ||
| AB158403.1 | - | 3 - 4 | 398 | 132 | ||
| BC127960.1 | - | 3 - 4 | 361 | 40134813 | 132 | |
| DV651301.1 | - | 2 - 3 | 237 | 132 | ||
| DV660756.1 | - | 4 - 5 | 482 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rph3al
Genomic Map