Assay Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029548.4 | NP_083824.1 | 6 - 7 | 757 | 59 | |
| GenBank mRNA | AB112925.1 | - | 6 - 7 | 724 | 59 | |
| AB158403.1 | - | 6 - 7 | 765 | 59 | ||
| BC127960.1 | - | 6 - 7 | 728 | 40134813 | 59 | |
| DV651301.1 | - | 5 - 6 | 604 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rph3al
Genomic Map