Assay Details
| Gene Symbol: Pdzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133226.2 | NP_573489.2 | 4 - 5 | 339 | 76 | |
| GenBank mRNA | AF334612.1 | - | 4 - 5 | 335 | 76 | |
| AK052748.1 | - | 4 - 5 | 340 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdzd3
Genomic Map