Assay Details
Gene Symbol: 4933416E03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040498.1 | 3 - 4 | 432 | 73 | ||
GenBank mRNA | AK016829.1 | - | 3 - 4 | 434 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAH (Asp-Glu-Ala-His) box polypeptide 35 | Dhx35 |
1200009D07Rik, Ddx35 |
-1.1mb | Sense | Chr. 2 - 158794770 - 158858220 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 9130015L21 gene | 9130015L21Rik | 42.3kb | Sense | Chr. 2 - 160016653 - 160040620 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
