Assay Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163504.1 | NP_001156976.1 | 1 - 2 | 220 | 71 | |
| GenBank mRNA | U09417.1 | - | 1 - 2 | 183 | 71 | |
| U09418.1 | - | 1 - 2 | 180 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163504.1, U09417.1, U09418.1 transcripts.
Set Membership: |
|

Nr1h4
Genomic Map