Assay Details
Gene Symbol: Mmp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013599.3 | NP_038627.1 | 1 - 2 | 181 | 78 | |
GenBank mRNA | AK004651.1 | - | 1 - 2 | 162 | 78 | |
AK142787.1 | - | 1 - 2 | 163 | 78 | ||
AK159292.1 | - | 1 - 2 | 163 | 78 | ||
AK161176.1 | - | 1 - 2 | 162 | 78 | ||
BC046991.1 | - | 1 - 2 | 149 | 6309245 | 78 | |
D12712.1 | - | 1 - 2 | 170 | 78 | ||
S67830.1 | - | 1 - 2 | 190 | 78 | ||
Z27231.1 | - | 1 - 2 | 162 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
