Assay Details
| Gene Symbol: Rfx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024918.1 | NP_001020089.1 | 16 - 17 | 2221 | 70 | |
| NM_027689.3 | NP_081965.2 | 12 - 13 | 1553 | 70 | ||
| GenBank mRNA | AB086957.1 | - | 16 - 17 | 2075 | 70 | |
| AB089184.1 | - | 12 - 13 | 1554 | 70 | ||
| AK034131.1 | - | 16 - 17 | 2208 | 70 | ||
| AK132673.1 | - | 13 - 14 | 1616 | 70 | ||
| AY102010.2 | - | 16 - 17 | 2104 | 70 | ||
| AY342003.1 | - | 16 - 17 | 2086 | 70 | ||
| BC138719.1 | - | 12 - 13 | 1541 | 8861741 | 70 | |
| BC138720.1 | - | 12 - 13 | 1541 | 8861742 | 70 | |
More Information
Set Membership: |
|

Rfx4
Genomic Map