| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000007713.mm.1 |
NM_001024918.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007713.mm.1 |
NM_001024918.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007713.mm.1 |
TC1000001024.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007718.mm.1 |
NM_001024918.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007718.mm.1 |
NM_001024918.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007718.mm.1 |
TC1000001024.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007714.mm.1 |
NM_001024918.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007714.mm.1 |
NM_001024918.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007714.mm.1 |
TC1000001024.mm.1 |
MTA TC |