Assay Details
| Gene Symbol: Rfx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024918.1 | NP_001020089.1 | 6 - 7 | 1013 | 85 | |
| NM_027689.3 | NP_081965.2 | 2 - 3 | 345 | 85 | ||
| GenBank mRNA | AB086957.1 | - | 6 - 7 | 867 | 85 | |
| AB089184.1 | - | 2 - 3 | 346 | 85 | ||
| AK016791.1 | - | 2 - 3 | 346 | 85 | ||
| AK034131.1 | - | 6 - 7 | 1000 | 85 | ||
| AK132673.1 | - | 3 - 4 | 408 | 85 | ||
| AY102010.2 | - | 6 - 7 | 896 | 85 | ||
| AY342003.1 | - | 6 - 7 | 878 | 85 | ||
| BC138719.1 | - | 2 - 3 | 333 | 8861741 | 85 | |
| BC138720.1 | - | 2 - 3 | 333 | 8861742 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx4
Genomic Map