Assay Details
| Gene Symbol: Rfx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024918.1 | NP_001020089.1 | 7 - 8 | 1090 | 128 | |
| NM_027689.3 | NP_081965.2 | 3 - 4 | 422 | 128 | ||
| GenBank mRNA | AB086957.1 | - | 7 - 8 | 944 | 128 | |
| AB089184.1 | - | 3 - 4 | 423 | 128 | ||
| AK034131.1 | - | 7 - 8 | 1077 | 128 | ||
| AK132673.1 | - | 4 - 5 | 485 | 128 | ||
| AY102010.2 | - | 7 - 8 | 973 | 128 | ||
| AY342003.1 | - | 7 - 8 | 955 | 128 | ||
| BC138719.1 | - | 3 - 4 | 410 | 8861741 | 128 | |
| BC138720.1 | - | 3 - 4 | 410 | 8861742 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx4
Genomic Map