Assay Details
| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291168.1 | NP_001278097.1 | 9 - 10 | 2787 | 78 | |
| NM_001291170.1 | NP_001278099.1 | 9 - 10 | 2267 | 78 | ||
| NM_001291173.1 | NP_001278102.1 | 8 - 9 | 1751 | 78 | ||
| NM_001291177.1 | NP_001278106.1 | 9 - 10 | 2382 | 78 | ||
| NM_001291178.1 | NP_001278107.1 | 8 - 9 | 1866 | 78 | ||
| NM_010899.3 | NP_035029.2 | 9 - 10 | 2902 | 78 | ||
| GenBank mRNA | EU887587.1 | - | 9 - 10 | 2668 | 78 | |
| EU887588.1 | - | 9 - 10 | 2728 | 78 | ||
| EU887589.1 | - | 9 - 10 | 2265 | 78 | ||
| EU887590.1 | - | 9 - 10 | 2365 | 78 | ||
| EU887591.1 | - | 8 - 9 | 1749 | 78 | ||
| EU887592.1 | - | 8 - 9 | 1849 | 78 | ||
| U36576.1 | - | 9 - 10 | 2798 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc2
Genomic Map