Assay Details
| Gene Symbol: Sall4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049188.1 | - | 3 - 4 | 2869 | 66 | |
| AK145388.1 | - | 3 - 4 | 2888 | 66 | ||
| AY463371.1 | - | 3 - 4 | 1437 | 66 | ||
| AY463372.1 | - | 2 - 3 | 402 | 66 | ||
| AY463373.1 | - | 3 - 4 | 2769 | 66 | ||
| BC099928.1 | - | 4 - 5 | 2555 | 66 | ||
More Information
Set Membership: |
|

Sall4
Genomic Map