Assay Details
| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011943.2 | NP_036073.1 | 1 - 2 | 310 | 129 | |
| GenBank mRNA | AK043482.1 | - | 1 - 2 | 311 | 129 | |
| AK077712.1 | - | 1 - 2 | 305 | 129 | ||
| AK086722.1 | - | 311 | 129 | |||
| AK138757.1 | - | 1 - 2 | 311 | 129 | ||
| AK141708.1 | - | 1 - 2 | 295 | 129 | ||
| AK162671.1 | - | 1 - 2 | 264 | 129 | ||
| BC075652.1 | - | 1 - 2 | 309 | 30541969 | 129 | |
| U39066.1 | - | 1 - 2 | 224 | 129 | ||
| X97052.1 | - | 1 - 2 | 168 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK043482.1, AK077712.1, AK086722.1, AK138757.1, AK141708.1, AK162671.1, BC075652.1, NM_011943.2, U39066.1, X97052.1 transcripts.
Set Membership: |
|

Map2k6
Genomic Map